4-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid ≥95%

Supplier: Matrix Scientific

043051-1G
101858-894EA 173.65 USD
101858-894
4-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid ≥95%
4-Methyl-2-(1-oxo-1,3-dihydro-2H-isoindol-2-yl)pentanoic acid

Hazard: Irritant

Formula: C₁₄H₁₇NO₃
MW: 247.3 g/mol
Melting Pt: 185…188 °C
MDL Number: MFCD02082683
CAS Number: 298700-70-6

Order Now

Specification Test Results

Melting point 185-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR