6,7-Dichloro-3-(trifluoromethyl)quinoxaline-2-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6,7-Dichloro-2-hydroxy-3-(trifluoromethyl)quinoxaline

043038-1G
101858-838EA 350.77 USD
101858-838
6,7-Dichloro-3-(trifluoromethyl)quinoxaline-2-one ≥95%
6,7-Dichloro-3-(trifluoromethyl)quinoxaline-2-one

Hazard: Irritant

Formula: C₉H₃Cl₂F₃N₂O
Melting Pt: 242…244 °C
MDL Number: MFCD09801051
CAS Number: 477857-25-3

Order Now

Specification Test Results

Melting point 242-244°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR