2-(4-Methoxyphenoxy)benzenecarbonitrile ≥95%
Supplier: Matrix Scientific
043037-500MG
043037-5G
101858-834EA
806.15
USD
101858-834
101858-830
2-(4-Methoxyphenoxy)benzenecarbonitrile ≥95%
2-(4-Methoxyphenoxy)benzenecarbonitrile
Hazard: Irritant
Formula:
C₁₄H₁₁NO₂ Melting Pt: 84…86 °C |
MDL Number:
MFCD02082665 CAS Number: 171771-88-3 |
Specification Test Results
Melting point | 84-86°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...