5-tert-Butyl-2-hydroxyisophthalaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(tert-Butyl)-2-hydroxyisophthalaldehyde

043031-1G
101858-802EA 350.77 USD
101858-802
5-tert-Butyl-2-hydroxyisophthalaldehyde ≥95%
5-tert-Butyl-2-hydroxyisophthalaldehyde

Hazard: Irritant

Formula: C₁₂H₁₄O₃
MW: 206.2400 g/mol
Melting Pt: 107 °C
MDL Number: MFCD01567135
CAS Number: 84501-28-0

Order Now

Specification Test Results

Melting point 107°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR