1-(4-Nitrobenzoyl)-4-piperidinecarboxamide ≥95%

Supplier: Matrix Scientific

043029-1G
101858-794EA 350.77 USD
101858-794
1-(4-Nitrobenzoyl)-4-piperidinecarboxamide ≥95%
1-(4-Nitrobenzoyl)-4-piperidinecarboxamide

Hazard: Irritant

Formula: C₁₃H₁₅N₃O₄
Melting Pt: 207…210 °C
MDL Number: MFCD00861248
CAS Number: 420812-64-2

Order Now

Specification Test Results

Melting point 207-210°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR