1-(4-Nitrobenzoyl)-4-piperidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

043028-500MG 043028-5G
101858-786EA 181.54 USD
101858-786 101858-790
1-(4-Nitrobenzoyl)-4-piperidinecarboxylic acid ≥95%
1-(4-Nitrobenzoyl)-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₅
Melting Pt: 202…204 °C
MDL Number: MFCD01568757
CAS Number: 303994-58-3

Order Now

Specification Test Results

Melting point 202-204°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR