4-[4-(Methoxycarbonyl)piperidino]-4-oxobutanoic acid ≥95%

Supplier: Matrix Scientific

043027-1G
101858-782EA 177.1 USD
101858-782
4-[4-(Methoxycarbonyl)piperidino]-4-oxobutanoic acid ≥95%
4-[4-(Methoxycarbonyl)piperidino]-4-oxobutanoic acid

Hazard: Irritant

Formula: C₁₁H₁₇NO₅
Melting Pt: 91…93 °C
MDL Number: MFCD01568756
CAS Number: 303994-57-2

Order Now

Specification Test Results

Melting point 91-93°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR