5-(Benzylsulfanyl)-1,3,4-thiadiazole-2-thiol ≥95%

Supplier: Matrix Scientific

Synonyms: 5-(Benzylthio)-1,3,4-thiadiazole-2-thiol

043022-5G 043022-500MG
101858-758EA 602.6 USD
101858-758 101858-754
5-(Benzylsulfanyl)-1,3,4-thiadiazole-2-thiol ≥95%
5-(Benzylsulfanyl)-1,3,4-thiadiazole-2-thiol

Hazard: Irritant

Formula: C₉H₈N₂S₃
MW: 240.37 g/mol
Melting Pt: 137…139 °C
MDL Number: MFCD01568740
CAS Number: 4858-36-0

Order Now

Specification Test Results

Melting point 137-139°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR