4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

043021-1G
101858-750EA 467.69 USD
101858-750
4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid ≥95%
4-Methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
Melting Pt: >256 °C (dec.)
MDL Number: MFCD01569174
CAS Number: 272437-91-9

Order Now

Specification Test Results

Melting point 256°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR