Methyl-4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate ≥95%
Supplier: Matrix Scientific
043020-500MG
043020-5G
101858-746EA
1073.85
USD
101858-746
101858-742
Methyl-4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate ≥95%
Methyl-4-methyl-3-oxo-3,4-dihydro-2H-1,4-benzothiazine-6-carboxylate
Hazard: Irritant
Formula:
C₁₁H₁₁NO₃S MW: 237.28 g/mol Melting Pt: 139…141 °C |
MDL Number:
MFCD01569173 CAS Number: 303987-90-8 |
Specification Test Results
Melting point | 139-141°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...