3-(2-Phenoxyphenyl)acrylic acid ≥95%
Supplier: Matrix Scientific
043013-1G
101858-706EA
698.46
USD
101858-706
3-(2-Phenoxyphenyl)acrylic acid ≥95%
3-(2-Phenoxyphenyl)acrylic acid
Hazard: Irritant
Formula:
C₁₅H₁₂O₃ Melting Pt: 182…184 °C |
MDL Number:
MFCD01568728 CAS Number: 95433-16-2 |
Specification Test Results
Melting point | 182-184°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...