4-(4-Fluorophenyl)-2-pyrimidinethiol ≥95%

Supplier: Matrix Scientific

043008-1G
101858-690EA 262.2 USD
101858-690
4-(4-Fluorophenyl)-2-pyrimidinethiol ≥95%
4-(4-Fluorophenyl)-2-pyrimidinethiol

Hazard: Irritant

Formula: C₁₀H₇FN₂S
Melting Pt: 225 °C
MDL Number: MFCD01315002
CAS Number: 155957-43-0

Order Now

Specification Test Results

Melting point 225°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR