1-[3-(4-Chlorophenyl)-5-isoxazolyl]-1-ethanol ≥95%

Supplier: Matrix Scientific

042998-1G
101858-638EA 467.69 USD
101858-638
1-[3-(4-Chlorophenyl)-5-isoxazolyl]-1-ethanol ≥95%
1-[3-(4-Chlorophenyl)-5-isoxazolyl]-1-ethanol

Hazard: Irritant

Formula: C₁₁H₁₀ClNO₂
Melting Pt: 63…67 °C
MDL Number: MFCD03425717
CAS Number: 109572-24-9

Order Now

Specification Test Results

Melting point 63-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR