1-(4-Methylbenzyl)-1H-indole-2,3-dione ≥95%

Supplier: Matrix Scientific

Synonyms: 1-[(4-Methylphenyl)methyl]-2,3-dihydro-1H-indole-2,3-dione

042997-500MG 042997-5G
101858-634EA 397.9 USD
101858-634 101858-630
1-(4-Methylbenzyl)-1H-indole-2,3-dione ≥95%
1-(4-Methylbenzyl)-1H-indole-2,3-dione

Hazard: Irritant

Formula: C₁₆H₁₃NO₂
MW: 251.29 g/mol
Melting Pt: 144…146 °C
MDL Number: MFCD00141570
CAS Number: 79183-26-9

Order Now

Specification Test Results

Melting point 144-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR