Ethyl-2-(5-amino-2-chloro-4-fluorophenoxy)acetate ≥95%

Supplier: Matrix Scientific

042994-1G
101858-626EA 927.69 USD
101858-626
Ethyl-2-(5-amino-2-chloro-4-fluorophenoxy)acetate ≥95%
Ethyl-2-(5-amino-2-chloro-4-fluorophenoxy)acetate

Hazard: Irritant

Formula: C₁₀H₁₁ClFNO₃
MW: 247.66 g/mol
Melting Pt: 76…80 °C
MDL Number: MFCD03425716
CAS Number: 91920-52-4

Order Now

Specification Test Results

Melting point 76-80°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR