[3-(2-Chlorophenyl)-5-isoxazolyl]methanol ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(2-Chlorophenyl)-5-(hydroxymethyl)isoxazole
, 3-(2-Chlorophenyl)-5-isoxazolemethanol
042991-1G
101858-610EA
467.69
USD
101858-610
[3-(2-Chlorophenyl)-5-isoxazolyl]methanol ≥95%
[3-(2-Chlorophenyl)-5-isoxazolyl]methanol
Hazard: Irritant
Formula:
C₁₀H₈ClNO₂ MW: 209.6300 g/mol Melting Pt: 73…76 °C |
MDL Number:
MFCD04124289 CAS Number: 438565-33-4 |
Specification Test Results
Melting point | 73-76°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...