2-Chloro-4-fluoro-5-nitrophenol ≥95%

Supplier: Matrix Scientific

042987-25G
101858-594EA 124.2 USD
101858-594
2-Chloro-4-fluoro-5-nitrophenol ≥95%
2-Chloro-4-fluoro-5-nitrophenol

Hazard: Irritant

Formula: C₆H₃ClFNO₃
MW: 191.5400 g/mol
Melting Pt: 105…108 °C
MDL Number: MFCD02670258
CAS Number: 84478-75-1

Order Now

Specification Test Results

Melting point 105-108°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR