N-Hydroxy-4-methoxybenzenecarboximidoyl chloride ≥95%

Supplier: Matrix Scientific

Synonyms: alpha-Chloro-4-methoxybenzaldoxime

042986-1G
101858-590EA 283.08 USD
101858-590
N-Hydroxy-4-methoxybenzenecarboximidoyl chloride ≥95%
N-Hydroxy-4-methoxybenzenecarboximidoyl chloride

Hazard: Irritant/Store in Freezer

Formula: C₈H₈ClNO₂
MW: 185.61 g/mol
Melting Pt: 87…89 °C
MDL Number: MFCD01924588
CAS Number: 38435-51-7

Order Now

Specification Test Results

Melting point 87-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR