4-(1-Methylhydrazino)-2-(trifluoromethyl)quinazoline ≥95%

Supplier: Matrix Scientific

042984-500MG
101858-578EA 921.54 USD
101858-578
4-(1-Methylhydrazino)-2-(trifluoromethyl)quinazoline ≥95%
4-(1-Methylhydrazino)-2-(trifluoromethyl)quinazoline

Hazard: Irritant

Formula: C₁₀H₉F₃N₄
MW: 242.21 g/mol
Melting Pt: 136…138 °C
MDL Number: MFCD00793700
CAS Number: 303148-88-1

Order Now

Specification Test Results

Melting point 136-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR