2-(1,2,3-Thiadiazol-4-yl)benzenol ≥95%

Supplier: Matrix Scientific

042983-500MG
101858-574EA 278.46 USD
101858-574
2-(1,2,3-Thiadiazol-4-yl)benzenol ≥95%
2-(1,2,3-Thiadiazol-4-yl)benzenol

Hazard: Irritant

Formula: C₈H₆N₂OS
Melting Pt: 111…113 °C
MDL Number: MFCD00792963
CAS Number: 59834-06-9

Order Now

Specification Test Results

Melting point 111-113°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR