(1-Amino-2,3-dihydro-1H-benzo[f]chromen-2-yl)methanol hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
{1-Amino-1H,2H,3H-naphtho[2,1-b]pyran-2-yl}methanol hydrochloride
042980-1G
101858-558EA
1400
USD
101858-558
(1-Amino-2,3-dihydro-1H-benzo[f]chromen-2-yl)methanol hydrochloride ≥95%
(1-Amino-2,3-dihydro-1H-benzo[f]chromen-2-yl)methanol hydrochloride
Hazard: Irritant
Formula:
C₁₄H₁₆ClNO₂ MW: 265.74 g/mol Melting Pt: 235…237 °C |
MDL Number:
MFCD00663795 CAS Number: 321392-00-1 |
Specification Test Results
Melting point | 235-237°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...