3-Amino-4-[(2,6-dichlorophenyl)sulfanyl]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

042976-1G
101858-538EA 192.31 USD
101858-538
3-Amino-4-[(2,6-dichlorophenyl)sulfanyl]benzenecarbonitrile ≥95%
3-Amino-4-[(2,6-dichlorophenyl)sulfanyl]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₃H₈Cl₂N₂S
Melting Pt: 180…184 °C
MDL Number: MFCD01935934
CAS Number: 338982-10-8

Order Now

Specification Test Results

Melting point 180-184°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR