3-Amino-4-[(4-chlorophenyl)sulfanyl]benzenecarbonitrile ≥95%

Supplier: Matrix Scientific

042974-1G
101858-526EA 467.69 USD
101858-526
3-Amino-4-[(4-chlorophenyl)sulfanyl]benzenecarbonitrile ≥95%
3-Amino-4-[(4-chlorophenyl)sulfanyl]benzenecarbonitrile

Hazard: Irritant

Formula: C₁₃H₉ClN₂S
MW: 260.75 g/mol
Melting Pt: 92…94 °C
MDL Number: MFCD00793340
CAS Number: 303147-30-0

Order Now

Specification Test Results

Melting point 92-94°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR