7-(4-Methoxystyryl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

042972-1G
101858-514EA 467.69 USD
101858-514
7-(4-Methoxystyryl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%
7-(4-Methoxystyryl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

Hazard: Irritant

Formula: C₁₄H₁₃N₅O
MW: 267.29 g/mol
Melting Pt: 255…257 °C
MDL Number: MFCD01935991
CAS Number: 303145-73-5

Order Now

Specification Test Results

Melting point 255-257°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR