7-(4-Methoxyphenyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%

Supplier: Matrix Scientific

042970-1G
101858-502EA 467.69 USD
101858-502
7-(4-Methoxyphenyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine ≥95%
7-(4-Methoxyphenyl)[1,2,4]triazolo[1,5-a]pyrimidin-2-amine

Hazard: Irritant

Formula: C₁₂H₁₁N₅O
MW: 241.25 g/mol
Melting Pt: 242…244 °C
MDL Number: MFCD00232449
CAS Number: 303145-64-4

Order Now

Specification Test Results

Melting point 242-244°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR