N-Allyl-3-chlorobenzenecarboxamide ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-N-(prop-2-en-1-yl)benzamide

042968-500MG
101858-490EA 181.54 USD
101858-490
N-Allyl-3-chlorobenzenecarboxamide ≥95%
N-Allyl-3-chlorobenzenecarboxamide

Hazard: Irritant

Formula: C₁₀H₁₀ClNO
MW: 195.65 g/mol
Melting Pt: 44…47 °C
MDL Number: MFCD01181345
CAS Number: 35306-52-6

Order Now

Specification Test Results

Melting point 44-47°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR