2-(5-Benzoyl-2-thienyl)acetonitrile ≥95%

Supplier: Matrix Scientific

042966-1G
101858-480EA 262.2 USD
101858-480
2-(5-Benzoyl-2-thienyl)acetonitrile ≥95%
2-(5-Benzoyl-2-thienyl)acetonitrile

Hazard: Irritant

Formula: C₁₃H₉NOS
Melting Pt: 122…124 °C
MDL Number: MFCD00664552
CAS Number: 303144-54-9

Order Now

Specification Test Results

Melting point 122-124°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR