6-Acetyl-7-methylpyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

042964-1G
101858-468EA 927.69 USD
101858-468
6-Acetyl-7-methylpyrazolo[1,5-a]pyrimidine-3-carbonitrile ≥95%
6-Acetyl-7-methylpyrazolo[1,5-a]pyrimidine-3-carbonitrile

Hazard: Irritant

Formula: C₁₀H₈N₄O
Melting Pt: 175…178 °C
MDL Number: MFCD00664254
CAS Number: 83702-52-7

Order Now

Specification Test Results

Melting point 175-178°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR