2-Cyano-3-(dimethylamino)-2-butenamide ≥97%
Supplier: Matrix Scientific
042959-1G
101858-444EA
698.46
USD
101858-444
2-Cyano-3-(dimethylamino)-2-butenamide ≥97%
2-Cyano-3-(dimethylamino)-2-butenamide
Hazard: Irritant
Formula:
C₇H₁₁N₃O Melting Pt: 194…196 °C |
MDL Number:
MFCD00663777 CAS Number: 62321-92-0 |
Specification Test Results
Melting point | 194-196°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...