3-(4-Fluoro-2-nitrophenoxy)-2-thiophenecarboxylic acid ≥95%
Supplier: Matrix Scientific
042957-1G
101858-432EA
350.77
USD
101858-432
3-(4-Fluoro-2-nitrophenoxy)-2-thiophenecarboxylic acid ≥95%
3-(4-Fluoro-2-nitrophenoxy)-2-thiophenecarboxylic acid
Hazard: Irritant
Formula:
C₁₁H₆FNO₅S MW: 283.24 g/mol Melting Pt: 159…161 °C |
MDL Number:
MFCD00214992 CAS Number: 303152-17-2 |
Specification Test Results
Melting point | 159-161°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...