4-Hydroxy-6-(3-methoxyphenyl)-2-phenyl-5-pyrimidinecarbonitrile ≥95%
Supplier: Matrix Scientific
042951-5G
042951-500MG
101858-400EA
1073.85
USD
101858-400
101858-396
4-Hydroxy-6-(3-methoxyphenyl)-2-phenyl-5-pyrimidinecarbonitrile ≥95%
4-Hydroxy-6-(3-methoxyphenyl)-2-phenyl-5-pyrimidinecarbonitrile
Hazard: Irritant
Formula:
C₁₈H₁₃N₃O₂ Melting Pt: 271…273 °C |
MDL Number:
MFCD00794095 CAS Number: 338982-08-4 |
Specification Test Results
Melting point | 271-273°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...