3-[(2-Amino-2-oxoethyl)sulfanyl]-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

042949-500MG
101858-392EA 558.9 USD
101858-392
3-[(2-Amino-2-oxoethyl)sulfanyl]-2-thiophenecarboxylic acid ≥95%
3-[(2-Amino-2-oxoethyl)sulfanyl]-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₇H₇NO₃S₂
Melting Pt: 231…235 °C
MDL Number: MFCD00663911
CAS Number: 303150-84-7

Order Now

Specification Test Results

Melting point 231-235°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR