4-(4-Methoxyphenyl)-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

042946-1G
101858-376EA 467.69 USD
101858-376
4-(4-Methoxyphenyl)-1H-pyrazol-5-amine ≥95%
4-(4-Methoxyphenyl)-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₁₀H₁₁N₃O
Melting Pt: 204…206 °C
MDL Number: MFCD00141526
CAS Number: 93439-79-3

Order Now

Specification Test Results

Melting point 204-206°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR