4-Chlorobenzenecarbaldehyde-N,N-dimethylhydrazone ≥95%

Supplier: Matrix Scientific

Synonyms: (E)-2-[(4-Chlorophenyl)methylidene]-1,1-dimethylhydrazine

042944-1G
101858-364EA 232.31 USD
101858-364
4-Chlorobenzenecarbaldehyde-N,N-dimethylhydrazone ≥95%
4-Chlorobenzenecarbaldehyde-N,N-dimethylhydrazone

Hazard: Irritant

Formula: C₉H₁₁ClN₂
MW: 182.65 g/mol
Melting Pt: 67…68 °C
MDL Number: MFCD01011933
CAS Number: 22699-29-2

Order Now

Specification Test Results

Melting point 67-68°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR