2-[Cyano(4-fluorophenyl)methyl]-6-fluorobenzenecarbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(2-Cyano-3-fluorophenyl)-2-(4-fluorophenyl)acetonitrile

042942-1G
101858-352EA 232.31 USD
101858-352
2-[Cyano(4-fluorophenyl)methyl]-6-fluorobenzenecarbonitrile ≥95%
2-[Cyano(4-fluorophenyl)methyl]-6-fluorobenzenecarbonitrile

Hazard: Irritant

Formula: C₁₅H₈F₂N₂
MW: 254.24 g/mol
Melting Pt: 72…75 °C
MDL Number: MFCD00231906
CAS Number: 303150-37-0

Order Now

Specification Test Results

Melting point 72-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR