2-(2-Anilino-1,3-thiazol-4-yl)benzenecarboxylic acid ≥95%

Supplier: Matrix Scientific

042936-500MG
101858-316EA 1108.6 USD
101858-316
2-(2-Anilino-1,3-thiazol-4-yl)benzenecarboxylic acid ≥95%
2-(2-Anilino-1,3-thiazol-4-yl)benzenecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₂N₂O₂S
Melting Pt: 228…231 °C
MDL Number: MFCD00141947
CAS Number: 303150-09-6

Order Now

Specification Test Results

Melting point 228-231°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR