2-(Allyloxy)-1-naphthaldehyde ≥95%

Supplier: Matrix Scientific

042932-1G
101858-296EA 115.38 USD
101858-296
2-(Allyloxy)-1-naphthaldehyde ≥95%
2-(Allyloxy)-1-naphthaldehyde

Hazard: Irritant

Formula: C₁₄H₁₂O
Melting Pt: 81…83 °C
MDL Number: MFCD00231645
CAS Number: 19530-43-9

Order Now

Specification Test Results

Melting point 81-83°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR