5-(3-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

042930-1G
101858-284EA 380 USD
101858-284
5-(3-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carbaldehyde ≥95%
5-(3-Chlorophenoxy)-1,3-dimethyl-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₁ClN₂O₂
Melting Pt: 59 °C
MDL Number: MFCD00215393
CAS Number: 109925-12-4

Order Now

Specification Test Results

Melting point 87-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR