1,3-Dimethyl-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde ≥95%

Supplier: Matrix Scientific

042929-1G
101858-280EA 1163.08 USD
101858-280
1,3-Dimethyl-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde ≥95%
1,3-Dimethyl-5-[3-(trifluoromethyl)phenoxy]-1H-pyrazole-4-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₁F₃N₂O₂
MW: 284.24 g/mol
Melting Pt: 84…86 °C
MDL Number: MFCD00215392
CAS Number: 109925-28-2

Order Now

Specification Test Results

Melting point 84-86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR