2-Methyl-2-(1H-1,2,4-triazol-1-yl)propanoic acid ≥95%
Supplier: Matrix Scientific
042925-1G
101858-258EA
410.77
USD
101858-258
2-Methyl-2-(1H-1,2,4-triazol-1-yl)propanoic acid ≥95%
2-Methyl-2-(1H-1,2,4-triazol-1-yl)propanoic acid
Hazard: Irritant
Formula:
C₆H₉N₃O₂ Melting Pt: 147…150 °C |
MDL Number:
MFCD00202110 CAS Number: 303148-48-3 |
Specification Test Results
Melting point | 147-150°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...