3-(Benzyloxy)-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

042923-1G
101858-246EA 192.31 USD
101858-246
3-(Benzyloxy)-2-thiophenecarboxylic acid ≥95%
3-(Benzyloxy)-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₀O₃S
Melting Pt: 123…125 °C
MDL Number: MFCD00214697
CAS Number: 186588-88-5

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR