1-Methyl-2,4-dioxohexahydro-5-pyrimidinecarboximidamide ≥95%
Supplier: Matrix Scientific
042921-1G
101858-234EA
1400
USD
101858-234
1-Methyl-2,4-dioxohexahydro-5-pyrimidinecarboximidamide ≥95%
1-Methyl-2,4-dioxohexahydro-5-pyrimidinecarboximidamide
Hazard: Irritant
Formula:
C₆H₁₀N₄O₂ Melting Pt: 275…277 °C |
MDL Number:
MFCD00172906 CAS Number: 446276-07-9 |
Specification Test Results
Melting point | 275-277°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...