3-Methyl-1-phenyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine ≥95%

Supplier: Matrix Scientific

042914-1G
101858-194EA 350.77 USD
101858-194
3-Methyl-1-phenyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine ≥95%
3-Methyl-1-phenyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₁₇H₁₄F₃N₃
Melting Pt: 104…105 °C
MDL Number: MFCD00140640
CAS Number: 321385-93-7

Order Now

Specification Test Results

Melting point 104-105°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR