2-Chloro-4-methyl-6,7-dihydro-5H-cyclopenta[b]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-methyl-5H,6H,7H-cyclopenta[b]pyridine

042908-500MG
101858-164EA 556.92 USD
101858-164
2-Chloro-4-methyl-6,7-dihydro-5H-cyclopenta[b]pyridine ≥95%
2-Chloro-4-methyl-6,7-dihydro-5H-cyclopenta[b]pyridine

Hazard: Irritant

Formula: C₉H₁₀ClN
MW: 167.64 g/mol
Melting Pt: 41…42 °C°C
MDL Number: MFCD08056605
CAS Number: 267242-99-9

Order Now

Specification Test Results

Melting point 58-62°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR