6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one

042907-1G
101858-160EA 201.54 USD
101858-160
6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one ≥95%
6-Chloro[1,2,4]triazolo[4,3-b]pyridazin-3(2H)-one

Hazard: Irritant

Formula: C₅H₃ClN₄O
MW: 170.56 g/mol
Melting Pt: 265…275 °C
MDL Number: MFCD01219639
CAS Number: 33050-32-7

Order Now

Specification Test Results

Melting point 265-275°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR