1-Oxo-2-(2-pyridinylmethyl)-1,2-dihydro-4-isoquinolinecarboxylic acid ≥95%

Supplier: Matrix Scientific

042905-500MG
101858-148EA 921.54 USD
101858-148
1-Oxo-2-(2-pyridinylmethyl)-1,2-dihydro-4-isoquinolinecarboxylic acid ≥95%
1-Oxo-2-(2-pyridinylmethyl)-1,2-dihydro-4-isoquinolinecarboxylic acid

Hazard: Irritant

Formula: C₁₆H₁₂N₂O₃
Melting Pt: 257 °C (dec.)
MDL Number: MFCD00140333
CAS Number: 477851-17-5

Order Now

Specification Test Results

Melting point 257°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR