Methyl-2-(4-chlorophenyl)-1-oxo-1,2-dihydro-4-isoquinolinecarboxylate ≥95%
Supplier: Matrix Scientific
042903-5G
042903-500MG
101858-136EA
747.69
USD
101858-136
101858-140
Methyl-2-(4-chlorophenyl)-1-oxo-1,2-dihydro-4-isoquinolinecarboxylate ≥95%
Methyl-2-(4-chlorophenyl)-1-oxo-1,2-dihydro-4-isoquinolinecarboxylate
Hazard: Irritant
Formula:
C₁₇H₁₂ClNO₃ Melting Pt: 152…154 °C |
MDL Number:
MFCD00140323 CAS Number: 446276-02-4 |
Specification Test Results
Melting point | 152-154°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...