Methyl-5-methyl-3-phenyl-4-isoxazolecarboxylate ≥95%

Supplier: Matrix Scientific

042902-1G
101858-132EA 349.6 USD
101858-132
Methyl-5-methyl-3-phenyl-4-isoxazolecarboxylate ≥95%
Methyl-5-methyl-3-phenyl-4-isoxazolecarboxylate

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MW: 217.23 g/mol
Melting Pt: 79…81 °C
MDL Number: MFCD00119105
CAS Number: 2065-28-3

Order Now

Specification Test Results

Melting point 79-81°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR