5-[4-(1H-Imidazol-1-yl)phenyl]-1H-pyrazole ≥95%

Supplier: Matrix Scientific

042896-1G
101858-100EA 349.6 USD
101858-100
5-[4-(1H-Imidazol-1-yl)phenyl]-1H-pyrazole ≥95%
5-[4-(1H-Imidazol-1-yl)phenyl]-1H-pyrazole

Hazard: Irritant

Formula: C₁₂H₁₀N₄
Melting Pt: 199…201 °C
MDL Number: MFCD00138185
CAS Number: 321385-58-4

Order Now

Specification Test Results

Melting point 199-201°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR