2-Chloro-5-nitro-1H-1,3-benzimidazole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-nitro-1H-benzo[d]imidazole

042895-5G
101858-096EA 384.62 USD
101858-096
2-Chloro-5-nitro-1H-1,3-benzimidazole ≥95%
2-Chloro-5-nitro-1H-1,3-benzimidazole

Hazard: Irritant

Formula: C₇H₄ClN₃O₂
MW: 197.58 g/mol
Melting Pt: 225…227 °C
MDL Number: MFCD00769824
CAS Number: 5955-72-6

Order Now

Specification Test Results

Melting point 225-227°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR